Skip to Content
MilliporeSigma
Adenosine 5′-diphosphate sodium salt bacterial, ≥95% (HPLC)

Adenosine 5′-diphosphate sodium salt

Adenosine 5′-diphosphate sodium salt bacterial, ≥95% (HPLC)
Synonym(s):
ADP
Empirical Formula (Hill Notation):
C10H15N5O10P2
CAS Number:
Molecular Weight:
427.20

Products

Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing