Skip to Content
MilliporeSigma
Acyclovir Related Compound F certified reference material, pharmaceutical secondary standard

Acyclovir Related Compound F

Acyclovir Related Compound F certified reference material, pharmaceutical secondary standard
Synonym(s):
N2-Acetylaciclovir, N-[9-[(2-Hydroxyethoxy)methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24

Products

Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade