Skip to Content
MilliporeSigma
α-Cyano-4-hydroxycinnamic acid suitable for matrix substance for MALDI-MS, Ultra pure

α-Cyano-4-hydroxycinnamic acid

α-Cyano-4-hydroxycinnamic acid suitable for matrix substance for MALDI-MS, Ultra pure
Synonym(s):
α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA
Linear Formula:
HOC6H4CH=C(CN)CO2H
CAS Number:
Molecular Weight:
189.17
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Analyte Suitability
Quality Segment
Functional Group
Markush Class
Markush Group
Technique