Skip to Content
MilliporeSigma
Actinonin

Actinonin

Actinonin
Synonym(s):
3-[[1-[(2-(Hydroxymethyl)-1-pyrrolidinyl)carbonyl]-2-methylpropyl]carbamoyl]octanohydroxamic acid
Empirical Formula (Hill Notation):
C19H35N3O5
CAS Number:
Molecular Weight:
385.50

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Activity Spectrum
Research Area
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing