Skip to Content
MilliporeSigma
Acid Red 33 analytical standard

Acid Red 33

Acid Red 33 analytical standard
Synonym(s):
5-Amino-4-hydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid disodium salt, Acid Red 33 disodium salt, Disodium 5-amino-4-hydroxy-3-(phenylazo)-naphthalene-2,7-disulfonate, Food Red 12
Empirical Formula (Hill Notation):
C16H11N3Na2O7S2
CAS Number:
Molecular Weight:
467.38
EC Number:

Products

Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing