Skip to Content
MilliporeSigma
Acetylcholine iodide ≥97%

Acetylcholine iodide

Acetylcholine iodide ≥97%
Synonym(s):
ACh, Acetylcholine iodide
Linear Formula:
(CH3)3N(I)CH2CH2OCOCH3
CAS Number:
Molecular Weight:
273.11

Products

Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing