Acetaminophen Related Compound A
Synonym(s):
4-(Acetylamino)phenyl acetate, p-Acetoxyacetanilide
Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2