Skip to Content
MilliporeSigma
Acesulfame potassium European Pharmacopoeia (EP) Reference Standard

Acesulfame potassium

Acesulfame potassium European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Acesulfame K, 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide potassium salt
Empirical Formula (Hill Notation):
C4H4KNO4S
CAS Number:
Molecular Weight:
201.24

Products

Product Category
Brand
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade