Skip to Content
MilliporeSigma
Acenaphthene certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Acenaphthene

Acenaphthene certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1,8-Ethylenenaphthalene, Acenaphthene
Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique