Skip to Content
MilliporeSigma
(−)-α-Bisabolol primary reference standard

(−)-α-Bisabolol

(−)-α-Bisabolol primary reference standard
Synonym(s):
(−)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol
Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique