Skip to Content
MilliporeSigma
α-[2-(Methylamino)ethyl]benzyl alcohol 97%

α-[2-(Methylamino)ethyl]benzyl alcohol

α-[2-(Methylamino)ethyl]benzyl alcohol 97%
Synonym(s):
3-(Methylamino)-1-phenylpropan-1-ol
Linear Formula:
C6H5CH(CH2CH2NHCH3)OH
CAS Number:
Molecular Weight:
165.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing