Skip to Content
MilliporeSigma
9-Fluorenemethanol 99%

9-Fluorenemethanol

9-Fluorenemethanol 99%
Synonym(s):
9-(Hydroxymethyl)fluorene, 9-Fluorenylmethanol
Empirical Formula (Hill Notation):
C14H12O
CAS Number:
Molecular Weight:
196.24
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing