Skip to Content
MilliporeSigma
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate ≥99%

9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate

9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate ≥99%
Synonym(s):
THA hydrochloride hydrate, Tacrine hydrochloride, Tetrahydroaminacrine hydrochloride hydrate
Empirical Formula (Hill Notation):
C13H14N2 · HCl · xH2O
CAS Number:
Molecular Weight:
234.72 (anhydrous basis)

Products

Product Category
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Markush Class
Markush Group