Skip to Content
MilliporeSigma
9,10-Bis(phenylethynyl)anthracene 97%

9,10-Bis(phenylethynyl)anthracene

9,10-Bis(phenylethynyl)anthracene 97%
Synonym(s):
BPEA
Empirical Formula (Hill Notation):
C30H18
CAS Number:
Molecular Weight:
378.46
EC Number:

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing