Skip to Content
MilliporeSigma
8-Quinolinol ≥99% (perchloric acid titration)

8-Quinolinol

8-Quinolinol ≥99% (perchloric acid titration)
Synonym(s):
8-Hydroxyquinoline, 8-Oxychinolin, Oxine
Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
Reference Material Type
Special Grade
Technique