Skip to Content
MilliporeSigma
8-Hydroxyjulolidine 96%

8-Hydroxyjulolidine

8-Hydroxyjulolidine 96%
Synonym(s):
2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol
Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing