Skip to Content
MilliporeSigma
8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate for fluorescence, ≥95% (TLC)

8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate

8-Anilino-1-naphthalenesulfonic acid hemimagnesium salt hydrate for fluorescence, ≥95% (TLC)
Synonym(s):
1,8-ANS magnesium salt, ANSA magnesium salt, Magnesium 8-anilino-1-naphthalenesulfonate hydrate, N-Phenyl peri acid magnesium salt
Empirical Formula (Hill Notation):
C16H12Mg0.5NO3S · xH2O
CAS Number:
Molecular Weight:
310.49 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application