Skip to Content
MilliporeSigma
8-Anilino-1-naphthalenesulfonic acid ammonium salt technical, ≥90% (NT)

8-Anilino-1-naphthalenesulfonic acid ammonium salt

8-Anilino-1-naphthalenesulfonic acid ammonium salt technical, ≥90% (NT)
Synonym(s):
1,8-ANS NH4, ANSA, Ammonium 8-anilino-1-naphthalenesulfonate, N-Phenyl peri acid
Linear Formula:
C16H13NO3S · NH3
CAS Number:
Molecular Weight:
316.37
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade