Skip to Content
MilliporeSigma
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 98%

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene 98%
Synonym(s):
1,3,4,6,7,8-Hexahydro-1-methyl-2H-pyrimido[1,2-a]pyrimidine, MTBD
Empirical Formula (Hill Notation):
C8H15N3
CAS Number:
Molecular Weight:
153.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing