Skip to Content
MilliporeSigma
7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt 10 mM in H2O

7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt

7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt 10 mM in H2O
Synonym(s):
−N7-dGTP, 7-Deaza-dGTP
Empirical Formula (Hill Notation):
C11H17N4O13P3
CAS Number:
Molecular Weight:
506.19

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Tissue
Showing 1-1 of 1
Compare
Product Number
Description
Pricing