Skip to Content
MilliporeSigma
7-Chloroisatin 97%

7-Chloroisatin

7-Chloroisatin 97%
Empirical Formula (Hill Notation):
C8H4ClNO2
CAS Number:
Molecular Weight:
181.58

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing