Skip to Content
MilliporeSigma
7-Benzyloxyindole 96%

7-Benzyloxyindole

7-Benzyloxyindole 96%
Synonym(s):
7-(Phenylmethoxy)indole, NSC 92526
Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing