Skip to Content
MilliporeSigma
6-(trifluoromethyl)pyridine-3-sulfonyl chloride 97%

6-(trifluoromethyl)pyridine-3-sulfonyl chloride

6-(trifluoromethyl)pyridine-3-sulfonyl chloride 97%
Empirical Formula (Hill Notation):
C6H3ClF3NO2S
CAS Number:
Molecular Weight:
245.61

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing