Skip to Content
MilliporeSigma
6-Phosphogluconic acid trisodium salt ≥95% anhydrous basis

6-Phosphogluconic acid trisodium salt

6-Phosphogluconic acid trisodium salt ≥95% anhydrous basis
Synonym(s):
D-Gluconate 6-phosphate trisodium salt
Empirical Formula (Hill Notation):
C6H10Na3O10P
CAS Number:
Molecular Weight:
342.08
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Application
Optical Isomer
Showing 1-2 of 2