Skip to Content
MilliporeSigma
6-Nitroveratraldehyde technical grade, 80%

6-Nitroveratraldehyde

6-Nitroveratraldehyde technical grade, 80%
Linear Formula:
O2NC6H2-3,4-(OCH3)2CHO
CAS Number:
Molecular Weight:
211.17
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing