Skip to Content
MilliporeSigma
6-Hydroxydopamine hydrochloride ≥97% (titration), powder

6-Hydroxydopamine hydrochloride

6-Hydroxydopamine hydrochloride ≥97% (titration), powder
Synonym(s):
2,4,5-Trihydroxyphenethylamine hydrochloride, 2,5-Dihydroxytyramine hydrochloride, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrochloride, 6-OHDA
Linear Formula:
(HO)3C6H2CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
205.64
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing