Skip to Content
MilliporeSigma
6-Bromohexanoyl chloride 97%

6-Bromohexanoyl chloride

6-Bromohexanoyl chloride 97%
Synonym(s):
6-Bromocaproyl chloride, 6-Bromohexanoic acid chloride
Linear Formula:
Br(CH2)5COCl
CAS Number:
Molecular Weight:
213.50
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing