Skip to Content
MilliporeSigma
6-Bromo-3-fluoro-2-formylpyridine AldrichCPR

6-Bromo-3-fluoro-2-formylpyridine

6-Bromo-3-fluoro-2-formylpyridine AldrichCPR
Empirical Formula (Hill Notation):
C6H3BrFNO
CAS Number:
Molecular Weight:
204.00

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing