Skip to Content
MilliporeSigma
6-Biopterin ≥97%

6-Biopterin

6-Biopterin ≥97%
Synonym(s):
(1’R,2’S)-Biopterin, 2-Amino-4-hydroxy-6-(1,2-dihydroxypropyl)pteridine, L-erythro-6-(1,2-Dihydroxypropyl)pterin
Empirical Formula (Hill Notation):
C9H11N5O3
CAS Number:
Molecular Weight:
237.22
EC Number:

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing