Skip to Content
MilliporeSigma
6-Aminofluorescein BioReagent, suitable for fluorescence, ~95% (HPLC)

6-Aminofluorescein

6-Aminofluorescein BioReagent, suitable for fluorescence, ~95% (HPLC)
Synonym(s):
Fluoresceinamine isomer II
Empirical Formula (Hill Notation):
C20H13NO5
CAS Number:
Molecular Weight:
347.32
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing