Skip to Content
MilliporeSigma
6′-Amino-D-luciferin ≥95.0% (HPLC)

6′-Amino-D-luciferin

6′-Amino-D-luciferin ≥95.0% (HPLC)
Synonym(s):
(S)-2-(6-Amino-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid, (S)-2-(6-Amino-2-benzothiazolyl)-4,5-dihydro-4-thiazolecarboxylic acid
Empirical Formula (Hill Notation):
C11H9N3O2S2
CAS Number:
Molecular Weight:
279.34

Products

Brand
Formula Weight
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing