Skip to Content
MilliporeSigma
6α-Hydroxycortisol ≥98%

6α-Hydroxycortisol

6α-Hydroxycortisol ≥98%
Synonym(s):
4-Pregnene-6α,11β,17,21-tetrol-3,20-dione
Empirical Formula (Hill Notation):
C21H30O6
CAS Number:
Molecular Weight:
378.46

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing