Skip to Content
MilliporeSigma
[6,6]-Phenyl C61 butyric acid methyl ester >99.5%

[6,6]-Phenyl C61 butyric acid methyl ester

[6,6]-Phenyl C61 butyric acid methyl ester >99.5%
Synonym(s):
1-[3-(Methoxycarbonyl)propyl]-1-phenyl-[6.6]C61, 3′H-Cyclopropa[1,9] [5,6]fullerene-C60-Ih-3′-butanoic acid 3′-phenyl methyl ester, PCBM, [60]PCBM
Empirical Formula (Hill Notation):
C72H14O2
CAS Number:
Molecular Weight:
910.88

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing