Skip to Content
MilliporeSigma
5-Methyl-2-hexanone 99%

5-Methyl-2-hexanone

5-Methyl-2-hexanone 99%
Synonym(s):
Isobutylacetone, Isopentyl methyl ketone, 5-Methyl-2-hexanone
Linear Formula:
(CH3)2CHCH2CH2COCH3
CAS Number:
Molecular Weight:
114.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing