Skip to Content
MilliporeSigma
5-Hydroxyindole-3-acetic acid analytical standard

5-Hydroxyindole-3-acetic acid

5-Hydroxyindole-3-acetic acid analytical standard
Synonym(s):
5-HIAA
Empirical Formula (Hill Notation):
C10H9NO3
CAS Number:
Molecular Weight:
191.18

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2