Skip to Content
MilliporeSigma
5-Fluoro-2-ethoxy-4(1H)pyrimidinone 97%

5-Fluoro-2-ethoxy-4(1H)pyrimidinone

5-Fluoro-2-ethoxy-4(1H)pyrimidinone 97%
Synonym(s):
2-Ethoxy-5-fluoropyrimidin-4(1H)-one
Linear Formula:
C4N2H2FOOCH2CH3
CAS Number:
Molecular Weight:
158.13
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing