Skip to Content
MilliporeSigma
5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide 95%

5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide

5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide 95%
Synonym(s):
4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonamide
Linear Formula:
ClC6H3(OCH3)CONHCH2C6H4SO2NH2
CAS Number:
Molecular Weight:
368.84
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing