Skip to Content
MilliporeSigma
5-Bromopyridine-3-carboxylic acid 98%

5-Bromopyridine-3-carboxylic acid

5-Bromopyridine-3-carboxylic acid 98%
Synonym(s):
5-Bromonicotinic acid
Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing