Skip to Content
MilliporeSigma
5-Bromo-4-chloro-3-indolyl sulfate potassium salt sulfatase substrate

5-Bromo-4-chloro-3-indolyl sulfate potassium salt

5-Bromo-4-chloro-3-indolyl sulfate potassium salt sulfatase substrate
Empirical Formula (Hill Notation):
C8H4BrClKNO4S
CAS Number:
Molecular Weight:
364.64

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1