Skip to Content
MilliporeSigma
5-Bromo-2-methyl-2-pentene 97%

5-Bromo-2-methyl-2-pentene

5-Bromo-2-methyl-2-pentene 97%
Linear Formula:
BrCH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
163.06

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing