![5-Bromo-2-methoxyaniline 95%](/deepweb/assets/sigmaaldrich/product/structures/316/888/3c7d439e-75a3-4a6b-92d9-9f534fbac37e/640/3c7d439e-75a3-4a6b-92d9-9f534fbac37e.png)
5-Bromo-2-methoxyaniline
![5-Bromo-2-methoxyaniline 95%](/deepweb/assets/sigmaaldrich/product/structures/316/888/3c7d439e-75a3-4a6b-92d9-9f534fbac37e/640/3c7d439e-75a3-4a6b-92d9-9f534fbac37e.png)
Synonym(s):
2-Amino-4-bromoanisole
Empirical Formula (Hill Notation):
C7H8BrNO
CAS Number:
Molecular Weight:
202.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing