Skip to Content
MilliporeSigma
5-Bromo-2-hydroxy-3-nitropyridine 98%

5-Bromo-2-hydroxy-3-nitropyridine

5-Bromo-2-hydroxy-3-nitropyridine 98%
Synonym(s):
5-Bromo-3-nitro-2(1H)-pyridinone, 5-Bromo-3-nitro-2-pyridinol
Empirical Formula (Hill Notation):
C5H3BrN2O3
CAS Number:
Molecular Weight:
218.99
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing