Skip to Content
MilliporeSigma
5-Bromo-2-chloropyrimidine 96%

5-Bromo-2-chloropyrimidine

5-Bromo-2-chloropyrimidine 96%
Empirical Formula (Hill Notation):
C4H2BrClN2
CAS Number:
Molecular Weight:
193.43

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing