Skip to Content
MilliporeSigma
5-(Benzyloxy)indole 95%

5-(Benzyloxy)indole

5-(Benzyloxy)indole 95%
Synonym(s):
NSC 62895
Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing