Skip to Content
MilliporeSigma
5-Amino-2-fluoropyridine 97%

5-Amino-2-fluoropyridine

5-Amino-2-fluoropyridine 97%
Synonym(s):
6-Fluoro-3-pyridineamine
Empirical Formula (Hill Notation):
C5H5FN2
CAS Number:
Molecular Weight:
112.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing