Skip to Content
MilliporeSigma
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Dye content 95 %

5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine

5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine Dye content 95 %
Empirical Formula (Hill Notation):
C80H90N8O8
CAS Number:
Molecular Weight:
1291.62

Products

Formula Weight
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing