Skip to Content
MilliporeSigma
5(6)-Carboxyfluorescein Dye content 90 %

5(6)-Carboxyfluorescein

5(6)-Carboxyfluorescein Dye content 90 %
Synonym(s):
5(6)-Carboxyfluorescein
Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Reagent Type