Skip to Content
MilliporeSigma
5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine 97%

5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine

5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine 97%
Synonym(s):
5,10,15,20-Tetra(4-pyridyl)porphyrin
Empirical Formula (Hill Notation):
C40H26N8
CAS Number:
Molecular Weight:
618.69
EC Number:

Products

Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing