Skip to Content
MilliporeSigma
4-Vinylbenzoic acid 97%

4-Vinylbenzoic acid

4-Vinylbenzoic acid 97%
Synonym(s):
Styrene-4-carboxylic acid, 4-Vinylbenzoic acid
Linear Formula:
H2C=CHC6H4CO2H
CAS Number:
Molecular Weight:
148.16

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing