Skip to Content
MilliporeSigma
4-(Trifluoromethyl)pyridine-3-carboxylic acid 98%

4-(Trifluoromethyl)pyridine-3-carboxylic acid

4-(Trifluoromethyl)pyridine-3-carboxylic acid 98%
Synonym(s):
4-(Trifluoromethyl)nicotinic acid
Empirical Formula (Hill Notation):
C7H4F3NO2
CAS Number:
Molecular Weight:
191.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing